Trimethyl orthobutyrate ≥97%
Supplier: Matrix Scientific
Synonyms:
Trimethylorthobutyrate, 1,1,1-Trimethoxybutane, Orthobutyric Acid Trimethyl Ester
086118-1G
086118-10G
086118-50G
102846-998EA
44.62
USD
102846-998
102847-000
102847-002
Trimethyl orthobutyrate ≥97%
Trimethyl orthobutyrate
Purity: 97%
CAS: 43083-12-1
MDL Number: MFCD00008480
Molecular Formula: C7H16O3
Molecular Weight: 148,2
Hazard: Irritant
Formula:
C₇H₁₆O₃ MW: 148.20 g/mol |
MDL Number:
MFCD00008480 CAS Number: 43083-12-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...