7-(2-Hydroxyethyl)theophylline ≥97%

Supplier: Matrix Scientific

Synonyms: 7-(2-Hydroxyethyl)-1,3-dimethyl-3,7-dihydro-1H-purine-2,6-dione, Etofylline

085943-1G 085943-10G
102846-442EA 72.31 USD
102846-442 102846-444
7-(2-Hydroxyethyl)theophylline ≥97%
7-(2-Hydroxyethyl)theophylline

Hazard: Irritant

Formula: C₉H₁₂N₄O₃
MW: 224.216 g/mol
MDL Number: MFCD00055055
CAS Number: 519-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR