3,5,5-Trimethylcyclohexane-1,2-dione ≥97%
Supplier: Matrix Scientific
085856-1G
085856-10G
085856-50G
102846-208EA
72.31
USD
102846-208
102846-210
102846-212
3,5,5-Trimethylcyclohexane-1,2-dione ≥97%
3,5,5-Trimethylcyclohexane-1,2-dione
Hazard: Irritant
Formula:
C₉H₁₄O₂ |
MDL Number:
MFCD02181023 CAS Number: 57696-89-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...