Indole ≥97%
Supplier: Matrix Scientific
Synonyms:
1H-Indole
085511-100G
085511-500G
102845-228EA
86.15
USD
102845-228
102845-230
Indole ≥97%
Indole
Hazard: Irritant
Formula:
C₈H₇N MW: 117.15 g/mol Flash Pt: 110 |
MDL Number:
MFCD00005607 CAS Number: 120-72-9 UN: 2811 ADR: 6.1 III Merck Index: 13,04985 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...