2-Vinylpyrazine ≥97%

Supplier: Matrix Scientific

085420-1G 085420-10G
102845-012EA 200 USD
102845-012 102845-014
2-Vinylpyrazine ≥97%
2-Vinylpyrazine

Hazard: Irritant

Formula: C₆H₆N₂
MDL Number: MFCD00051591
CAS Number: 4177-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR