Phosphonitrilic chloride trimer ≥97%
Supplier: Matrix Scientific
Synonyms:
Cyclic Phosphonitrilic Chloride Trimer, Hexachlorocyclotriphosphazene
084077-1G
084077-10G
084077-50G
102841-464EA
56.92
USD
102841-464
102841-466
102841-468
Phosphonitrilic chloride trimer ≥97%
Phosphonitrilic chloride trimer
Purity: 97%
CAS: 940-71-6
MDL Number: MFCD00006474
Molecular Formula: Cl6N3P3
Molecular Weight: 347,66
Hazard: Irritant
Formula:
Cl₆N₃P₃ |
MDL Number:
MFCD00006474 CAS Number: 940-71-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...