Piperonyloyl chloride ≥95%

Supplier: Matrix Scientific

Synonyms: 1,3-Benzodioxole-5-carbonyl chloride

083192-1G 083192-10G
102839-142EA 127.69 USD
102839-142 102839-144
Piperonyloyl chloride ≥95%
Piperonyloyl chloride

Hazard: Irritant

Formula: C₈H₅ClO₃
MW: 184.575 g/mol
MDL Number: MFCD00016904
CAS Number: 25054-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR