Neopentyl bromide ≥97%
Supplier: Matrix Scientific
Synonyms:
2,2-Dimethyl-1-propyl bromide, 1-Bromo-2,2-dimethylpropane
084168-1G
084168-10G
102841-700EA
72.31
USD
102841-700
102841-702
Neopentyl bromide ≥97%
Neopentyl bromide
Hazard: Flammable
Formula:
C₅H₁₁Br Boiling Pt: 104…106 °C Density: 1.095 Flash Pt: 6 °C |
MDL Number:
MFCD00000209 CAS Number: 630-17-1 UN: 1993 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...