4-Amino-3-chloro-5-methylbenzoic acid ≥97%
Supplier: Matrix Scientific
082216-1G
082216-10G
102836-836EA
353.85
USD
102836-836
102836-838
4-Amino-3-chloro-5-methylbenzoic acid ≥97%
4-Amino-3-chloro-5-methylbenzoic acid
Hazard: Irritant
Formula:
C₈H₈ClNO₂ |
MDL Number:
MFCD01313709 CAS Number: 157069-52-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...