2,6-Diaminopurine ≥97%

Supplier: Matrix Scientific

Synonyms: 9H-Purine-2,6-diamine, 7H-Purine-2,6-diamine

081455-10G 081455-50G
102834-978EA 310.77 USD
102834-978 102834-980
2,6-Diaminopurine ≥97%
2,6-Diaminopurine

Hazard: Irritant

Formula: C₅H₆N₆
MW: 150.14 g/mol
MDL Number: MFCD00213668
CAS Number: 1904-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR