Imidazolidin-4-one hydrochloride ≥95%

Supplier: Matrix Scientific

076581-250MG 076581-1G 076581-5G
102546-944EA 400 USD
102546-944 102546-946 102546-948
Imidazolidin-4-one hydrochloride ≥95%
Imidazolidin-4-one hydrochloride

Hazard: Irritant

Formula: C₃H₇ClN₂O
MDL Number: MFCD22194318
CAS Number: 1373253-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR