Isoxazolo[4,5-b]pyridin-3-amine ≥95%

Supplier: Matrix Scientific

076254-250MG 076254-1G 076254-5G
102545-230EA 427.69 USD
102545-230 102545-232 102545-234
Isoxazolo[4,5-b]pyridin-3-amine ≥95%
Isoxazolo[4,5-b]pyridin-3-amine

Hazard: Irritant

Formula: C₆H₅N₃O
MW: 135.1200 g/mol
MDL Number: MFCD13193314
CAS Number: 114080-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR