Pirfenidone ≥95%
Supplier: Matrix Scientific
076078-1G
076078-5G
102544-382EA
36.92
USD
102544-382
102544-384
Pirfenidone ≥95%
Pirfenidone
Purity: 95+%
CAS: 53179-13-8
MDL Number: MFCD00866047
Molecular Formula: C12H11NO
Molecular Weight: 185,23
Hazard: Irritant
Formula:
C₁₂H₁₁NO |
MDL Number:
MFCD00866047 CAS Number: 53179-13-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...