Quinidine ≥95%

Supplier: Matrix Scientific

Synonyms: (+)-Quinidine

075925-100G 075925-25G
102543-662EA 270.77 USD
102543-662 102833-890
Quinidine ≥95%
Quinidine

Hazard: Irritant

Formula: C20H24N2O2
MW: 324.42 g/mol
Melting Pt: 171…173 °C
MDL Number: MFCD00135581
CAS Number: 56-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR