(4-Nitrophenyl)acetonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Nitrobenzyl cyanide
075620-10G
075620-100G
102542-190EA
55.38
USD
102542-190
102542-192
(4-Nitrophenyl)acetonitrile ≥95%
(4-Nitrophenyl)acetonitrile
Hazard: Toxic
Formula:
C₈H₆N₂O₂ MW: 162.15 g/mol Boiling Pt: 336 Melting Pt: 114…117 °C |
MDL Number:
MFCD00007372 CAS Number: 555-21-5 UN: 3439 ADR: 6.1 III Merck Index: 12,06688 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...