D-(+)-Raffinose pentahydrate ≥95%

Supplier: Matrix Scientific

Synonyms: Gossypose pentahydrateMelitose pentahydrateO-α-D-Galactopyranosyl-(1→6)-α-D-glucopyranosyl β-D-fructofuranoside(3R,4S,5S,6R)-2-(((2S,3S,4S,5R)-3,4-Dihydroxy-2,5-bis(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-6-((((3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-3,4,5-triol pentahydrateD-(+)-Raffinose pentahydrateMelitriose pentahydrate

075364-100G
102541-020EA 138.46 USD
102541-020
D-(+)-Raffinose pentahydrate ≥95%
D(+)-Raffinose pentahydrate

Hazard: Irritant

Formula: C₁₈H₃₂O₁₆·5H₂O
MW: 594.52 g/mol
MDL Number: MFCD00071590
CAS Number: 17629-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR