L-Phenylalanine ≥95%

Supplier: Matrix Scientific

Synonyms: H-L-Phe-OH, Phenylalanine, H-Phe-OH, L-α-Phenylalanine, (S)-(-)-Phenylalanine, 3-Phenyl-L-alanine, 3-Phenyl-L(-)-alanine

075335-100G 075335-500G
102540-888EA 60 USD
102540-888 102540-890
L-Phenylalanine ≥95%
L-α-Phenylalanine

Hazard: Irritant

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Melting Pt: 227 °C. (dec.)
MDL Number: MFCD00064227
CAS Number: 63-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR