(R)-(-)-Methyl mandelate ≥95%

Supplier: Matrix Scientific

Synonyms: D(-)-Methyl glycolate, (R)-(-)-Methyl glycolate, (R)-(-)-Mandelic acid methyl ester, D(-)-Methyl mandelate, D(-)-2-Hydroxy-2-phenylacetic acid methyl ester, D(-)-Mandelic acid methyl ester, (R)-Methyl 2-hydroxy-2-phenylacetate, Methyl (R)-(-)-mandelate, (R)-(-)-2-Hydroxy-2-phenylacetic acid methyl ester

075258-5G 075258-10G 075258-100G
102540-534EA 90.77 USD
102540-534 102540-536 102540-538
(R)-(-)-Methyl mandelate ≥95%
(R)-(-)-Methyl mandelate

Hazard: Irritant

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 267
MDL Number: MFCD00064247
CAS Number: 20698-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR