N-Benzylglycine hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
Bzl-Gly-OH·HCl, Bzl-Gly-OH.HCl
075115-10G
075115-100G
102539-838EA
107.69
USD
102539-838
102539-840
N-Benzylglycine hydrochloride ≥95%
N-Benzylglycine hydrochloride
Hazard: Irritant
Formula:
C₉H₁₁NO₂·HCl MW: 201.7 g/mol |
MDL Number:
MFCD00156934 CAS Number: 7689-50-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...