D-(+)-Biotin ≥95%

Supplier: Matrix Scientific

Synonyms: Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7

074935-25G 074935-100G
102833-742EA 246.15 USD
102833-742 102833-744
D-(+)-Biotin ≥95%
D(+)-Biotin

TCA listed: T
Hazard: Irritant

Formula: C₁₀H₁₆N₂O₃S
MW: 244.31 g/mol
Melting Pt: 232 °C (dec.)
Density: 1.42
MDL Number: MFCD00005541
CAS Number: 58-85-5
Merck Index: 13,01231

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR