2-Bromethylamine hydrobromide ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromoethylammonium bromide, 2-Bromoethylamine hydrobromide

074880-100G 074880-500G
102538-664EA 61.54 USD
102538-664 102538-666
2-Bromethylamine hydrobromide ≥95%
2-Bromethylamine hydrobromide

Hazard: Irritant

Formula: BrCH₂CH₂NH₃Br
MW: 204.89 g/mol
MDL Number: MFCD00012886
CAS Number: 2576-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR