1-(p-Toluenesulfonyl)imidazole ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(P-Toluenesulfonyl)imidazole, 1-(P-Toluenesulphonyl)imidazole, 1-Tosylimidazole

074873-100G 074873-25G
102538-636EA 230.77 USD
102538-636 102833-734
1-(p-Toluenesulfonyl)imidazole ≥95%
1-(p-Toluenesulphonyl)imidazole

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₂S
MW: 222.26 g/mol
Melting Pt: 77…78 °C
MDL Number: MFCD00005285
CAS Number: 2232-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR