(S)-(+)-Camptothecin ≥95%
Supplier: Matrix Scientific
Synonyms:
(S)-4-Ethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, (+)-Camptothecin, 4-Ethyl-4-hydroxy-1H-pyrano-[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione
074699-1G
074699-5G
102537-764EA
76.92
USD
102537-764
102537-766
(S)-(+)-Camptothecin ≥95%
(S)-(+)-Camptothecin
Hazard: Irritant
Formula:
C₂₀H₁₆N₂O₄ MW: 348.36 g/mol Melting Pt: 255…257 °C (dec.) |
MDL Number:
MFCD00081076 CAS Number: 7689-03-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...