1-Benzyl-4-hydroxypiperidine ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Benzylpiperidin-4-ol

074252-5G 074252-100G
102535-450EA 72.31 USD
102535-450 102535-454
1-Benzyl-4-hydroxypiperidine ≥95%
1-Benzyl-4-hydroxypiperidine

Hazard: Irritant

Formula: C₁₂H₁₇NO
MDL Number: MFCD00006503
CAS Number: 4727-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR