6-Chloro-1H-indazole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

073554-1G 073554-5G
102530-784EA 569.23 USD
102530-784 102530-786
6-Chloro-1H-indazole-3-carboxylic acid ≥95%
6-Chloro-1H-indazole-3-carboxylic acid

Hazard: Irritant

Formula: C₈H₅ClN₂O₂
MDL Number: MFCD05663998
CAS Number: 129295-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR