6-Nitro-m-anisic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Methoxy-2-nitrobenzoic acid

073213-100G 073213-25G
102528-980EA 152.31 USD
102528-980 102663-938
6-Nitro-m-anisic acid ≥95%
6-Nitro-m-anisic acid

Hazard: Irritant

Formula: C₈H₇NO₅
MW: 197.15 g/mol
Melting Pt: 138
MDL Number: MFCD00151836
CAS Number: 1882-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR