3-Bromo-o-toluidine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-6-bromotoluene, 3-Bromo-2-methylaniline

073016-25G 073016-100G
102833-582EA 115.38 USD
102833-582 102833-584
3-Bromo-o-toluidine ≥95%
3-Bromo-o-toluidine

Hazard: Toxic/Light Sensitive

Formula: C₇H₈BrN
Boiling Pt: 253…255 °C
Density: 1.51
MDL Number: MFCD00051579
CAS Number: 55289-36-6
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR