2-Chloro-5-(trifluoromethyl)pyridine-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-5-(trifluoromethyl)isonicotinic acid

072995-1G 072995-5G 072995-10G
102527-828EA 84.62 USD
102527-828 102527-830 102527-832
2-Chloro-5-(trifluoromethyl)pyridine-4-carboxylic acid ≥95%
2-Chloro-5-(trifluoromethyl)pyridine-4-carboxylic acid

Hazard: Irritant

Formula: C₇H₃ClF₃NO₂
MDL Number: MFCD01862019
CAS Number: 505084-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR