7-Hydroxycoumarin ≥95%
Supplier: Matrix Scientific
Synonyms:
Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone
072969-10G
072969-100G
102527-706EA
30.77
USD
102527-706
102527-708
7-Hydroxycoumarin ≥95%
7-Hydroxycoumarin
Hazard: Irritant
Formula:
C₉H₆O₃ MW: 162.14 g/mol Boiling Pt: 382 |
MDL Number:
MFCD00006878 CAS Number: 93-35-6 Merck Index: 12,09981 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...