2,5-Dibromobenzoic acid ≥95%

Supplier: Matrix Scientific

072908-10G 072908-100G
102527-382EA 143.08 USD
102527-382 102527-384
2,5-Dibromobenzoic acid ≥95%
2,5-Dibromobenzoic acid

Hazard: Irritant

Formula: C₇H₄Br₂O₂
MDL Number: MFCD00016494
CAS Number: 610-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR