2-Bromo-3-nitrobenzoic acid ≥95%
Supplier: Matrix Scientific
072896-5G
072896-25G
102527-322EA
44.62
USD
102527-322
102551-816
2-Bromo-3-nitrobenzoic acid ≥95%
2-Bromo-3-nitrobenzoic acid
Hazard: Irritant
Formula:
C₇H₄BrNO₄ Melting Pt: 184…186 °C |
MDL Number:
MFCD00074899 CAS Number: 573-54-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...