2-Amino-6-chloro-3-nitropyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Chloro-3-nitro-2-pyridylamine, 6-Chloro-3-nitropyridin-2-amine

072792-100G 072792-25G
102526-772EA 184.62 USD
102526-772 102833-550
2-Amino-6-chloro-3-nitropyridine ≥95%
2-Amino-6-chloro-3-nitropyridine

Hazard: Irritant

Formula: C₅H₄ClN₃O₂
MDL Number: MFCD00044350
CAS Number: 27048-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR