2-chloro-5-iodopyrimidine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-5-iodopyrimidine

072113-1G 072113-5G
102523-162EA 89.23 USD
102523-162 102523-164
2-chloro-5-iodopyrimidine ≥95%
2-chloro-5-iodopyrimidine

Hazard: Irritant

Formula: C₄H₂ClIN₂
MDL Number: MFCD06200210
CAS Number: 32779-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR