[2-(4-Ethylpiperazin-1-yl)phenyl]methanol ≥95%

Supplier: Matrix Scientific

071845-1G 071845-5G 071845-10G
102521-670EA 284.62 USD
102521-670 102521-672 102521-674
[2-(4-Ethylpiperazin-1-yl)phenyl]methanol ≥95%
[2-(4-Ethylpiperazin-1-yl)phenyl]methanol

Hazard: Irritant

Formula: C₁₃H₂₀N₂O
CAS Number: 914349-49-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR