4-(4-Methylphenyl)-1,2-dihydrophthalazin-1-one ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(p-Tolyl)phthalazin-1(2H)-one

071366-500MG 071366-1G
102519-910EA 403.08 USD
102519-910 102519-912
4-(4-Methylphenyl)-1,2-dihydrophthalazin-1-one ≥97%
4-(4-Methylphenyl)-1,2-dihydrophthalazin-1-one

Hazard: Irritant

Formula: C₁₅H₁₂N₂O
MDL Number: MFCD00222976
CAS Number: 51334-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR