Pterostilbene ≥95%
Supplier: Matrix Scientific
Synonyms:
(E)-4-(3,5-Dimethoxystyryl)phenol, 4-[(E)-2-(3,5-Dimethoxyphenyl)ethenyl]phenol
063629-5G
063629-25G
102551-622EA
43.08
USD
102551-622
102551-624
Pterostilbene ≥95%
Pterostilbene
Hazard: Irritant
Formula:
C₁₆H₁₆O₃ MW: 256.2964 g/mol |
MDL Number:
MFCD00238710 CAS Number: 537-42-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...