3-Aminobutan-1-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-butan-1-ol

061407-1G 061407-5G
102518-888EA 15.38 USD
102518-888 102833-312
3-Aminobutan-1-ol ≥95%
3-Aminobutan-1-ol

Hazard: Irritant

Formula: C₄H₁₁NO
MDL Number: MFCD03412694
CAS Number: 2867-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR