Tropine
Supplier: Matrix Scientific
Synonyms:
endo-8-Methyl-8-azabicyclo[3.2.1]octan-3-ol
058452-25G
058452-100G
102551-534EA
81.54
USD
102551-534
102551-536
Tropine
Tropine
Hazard: Irritant
Formula:
C₈H₁₅NO MW: 141.21 g/mol |
MDL Number:
MFCD00180667 CAS Number: 120-29-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...