Chlorpromazine hydrochloride

Supplier: Matrix Scientific

Synonyms: 2-Chloro-10-[3-(dimethylamino)-1-propyl]phenothiazine Hydrochloride

058317-25G 058317-100G
102833-258EA 143.08 USD
102833-258 102833-260
Chlorpromazine hydrochloride
Chlorpromazine hydrochloride

CAS: 69-09-0 MDL Number: MFCD00012654 Molecular Formula: C17H20Cl2N2S Molecular Weight: 355,33 Hazard: Irritant

Formula: C₁₇H₁₉ClN₂S
MW: 355.32 g/mol
MDL Number: MFCD00012654
CAS Number: 69-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR