Piperazine

Supplier: Matrix Scientific

058316-100G 058316-500G
102551-522EA 61.54 USD
102551-522 102551-524
Piperazine
Piperazine

Hazard: Irritant

Formula: C₄H₁₀N₂
MW: 86.14 g/mol
MDL Number: MFCD00005953
CAS Number: 110-85-0
UN: 2579
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR