4-(tert-Butoxycarbonyl)phenylboronic acid ≥95%

Supplier: Matrix Scientific

057724-1G 057724-5G
102518-856EA 44.62 USD
102518-856 102518-858
4-(tert-Butoxycarbonyl)phenylboronic acid ≥95%
4-(tert-Butoxycarbonyl)phenylboronic acid

Hazard: Irritant/Refrigerate/Stored under Argon

Formula: C₁₁H₁₅BO₄
Melting Pt: 100…106 °C
MDL Number: MFCD03411946
CAS Number: 850568-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR