Melamine, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.08614.9025
EM8.08614.9025EA 835 USD
EM8.08614.9025
Melamine, Sigma-Aldrich®
Melamine
Formula: C₃H₆N₆
Density: 1.57 g/cm³ (20 °C)
Flash Pt: >280 °C
MDL Number: MFCD00006055
CAS Number: 108-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR