Big Mouth Step Screw-Thread Vials, Restek
Supplier: Restek
24629 24626 24627 24628
RK24629PK 362.69 USD
RK24629 10854-344 10854-346 10854-348
Big Mouth Step Screw-Thread Vials, Restek
Vials
Big mouth step screw-thread vials feature a 2 ml volume and a 10 mm mouth.
  • 10 mm orifice
  • Screw-thread format
  • White graduated markings
  • Clear or amber color
These vials have a 2 ml capacity with obvious white marking spots.