Renal Artery Clamp Cooley, OR Grade, Sklar®
Supplier: Sklar
52-6965 52-6966 52-6968
10197-098EA 880.63 USD
10197-098 10197-100 10197-102
Renal Artery Clamp Cooley, OR Grade, Sklar®
Forceps
Atraumatic Renal Artery Clamp is a ratcheted, finger ring clamp used in cardiothoracic procedures for controlling blood flow in the renal vessels.
  • Made of stainless steel
  • Blunt tip configuration
  • Cause little to no damage to the vessel walls
  • These are angled twice to create a basket shape with 2×2 rows of fine, atraumatic teeth