Econo™ Sterile Littauer Suture Scissors, Sklar
Supplier: Sklar
96-2512 96-2513 96-2662
89023-776CS 355.99 USD
89023-776 89023-778 89023-982
Econo™ Sterile Littauer Suture Scissors, Sklar
Scissors
Econo™ Sterile Littauer Suture Scissors are a ready-to-use, sterile, disposable, floor-grade instrument used for suture removal
These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. Littauer suture scissors typically have larger, less delicate tips than Spencer Stitch Scissors, and are often used for removing larger sutures. They should not be used for cutting tissue.