Human Dermal Fibroblasts, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
Lifeline® Normal Human Dermal Fibroblasts are cryopreserved as primary cells to ensure the highest viability and plating efficiency.
- Plating efficiency
- Quality tested to ensure optimal growth
- No exposure to antimicrobials or phenol red
- Ensures proper growth and morphology
Lifeline® Normal Human Dermal Fibroblasts Neonatal are isolated from neonatal human foreskin and cryopreserved as Primary cells to ensure the highest viability and plating efficiency
Our skin fibroblasts are quality tested in FibroLife® S2 (2% serum) or FibroLife® Xeno-Free Medium to ensure optimal growth at rates equal to or greater than classical serum-supplemented medium, and to ensure proper growth and morphology over a period of at least 15 population doublings.
Lifeline® dermal fibroblasts are not exposed to antimicrobials or phenol red.
Lifeline® Normal Human Dermal Fibroblasts Adult are isolated from adult human skin and cryopreserved as Primary cells to ensure the highest viability and plating efficiency.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)