Love-Gruenwald Intervertebral Disc Rongeur, OR Grade, Sklar®
Supplier: Sklar
42-3150 42-3152 42-3156 42-3158 42-3160
10196-950EA 849.59 USD
10196-950 10196-952 10196-954 10196-956 10196-958
Love-Gruenwald Intervertebral Disc Rongeur, OR Grade, Sklar®
Bone Cutters and Rongeurs
Love-Gruenwald intervertebral disc rongeurs are made of stainless steel.
  • With industrial standard
  • OR-grade
They are used to cut and remove bone fragments in the spinal column, and are available in a variety of lengths and cut patterns.