MEF Feeder Cells, Cell Biolabs
Supplier: Cell Biolabs
CBA-310 CBA-312
102966-510EA 387.69 USD
102966-510 102966-514
MEF Feeder Cells, Cell Biolabs
Cells
MEF Feeder Cells are mouse embroynic fibrobasts that have been traditionally used for years to promote stem cell culture
Cell Biolabs' MEF cells are not primary cells and can typically be passaged approximately 6 weeks without becoming senescent. Available with or without an antiobiotic resistance marker.