Ward's® Preserved Dogfish Sharks, 22 to 27"
Supplier: Avantor
132.1095.1 132.1115.1 132.1135.1
470000-372PK 196.99 USD
470000-372 470000-374 470000-376 470000-378 470001-474 470001-478 470001-480
Ward's® Preserved Dogfish Sharks, 22 to 27"
Specimens Preserved Specimens Preserved Animal Materials
Standard formalin preserved dogfish sharks.
  • Cartilaginous fish anatomy
  • No swim bladder or operculum
  • Gill slits
  • Good for group dissection
Dogfish sharks, Squalus sp., are a popular teaching specimen for biology and anatomy courses, specifically used for the teaching of cartilaginous fish anatomy. They are also great animal models to study comparative and vertebrate anatomy.
Dogfish will either be species Squalus sp. or Mustelus canis.
The harvesting of dogfish sharks is regulated by National Marine Fisheries to protect the species. Quantities used by our industry are very small in relation to the overall harvest.
Packaging: Packed in pails or vacuum-sealled bags.