OxiSelect™ 96-Well Comet Assay Kits and Slides, Cell Biolabs
Supplier: Cell Biolabs
STA-355 STA-355-5 STA-356 STA-356-5
102964-992EA 455.38 USD
102964-992 102964-994 102964-996 102964-998
OxiSelect™ 96-Well Comet Assay Kits and Slides, Cell Biolabs
Assays Cellular Assays
OxiSelect™ 96-Well Comet Assay Kits and Slides provide a fast, convenient way to screen for general DNA damage, regardless of the source or nature of the damage
Kits include Comet Slides, reagents, and a fluorescent dye to visualize cells under an epifluorescence microscope. Slides are available to order separately.